The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001165937.1 | NP_001159409.1 | 11 - 12 | 1155 | 71 |
NM_001165938.1 | NP_001159410.1 | 10 - 11 | 1101 | 71 | |
NM_006804.3 | NP_006795.3 | 11 - 12 | 1155 | 71 | |
XM_006721646.2 | XP_006721709.1 | 10 - 11 | 1025 | 71 | |
XM_011524210.1 | XP_011522512.1 | 11 - 12 | 1124 | 71 | |
XM_011524211.2 | XP_011522513.1 | 11 - 12 | 1127 | 71 | |
XM_011524213.1 | XP_011522515.1 | 10 - 11 | 1025 | 71 | |
XM_017024041.1 | XP_016879530.1 | 11 - 12 | 1127 | 71 | |
GenBank mRNA | AB209772.1 | - | 3 - 4 | 1495 | 71 |
AK123364.1 | - | 2 - 3 | 1182 | 71 | |
AK225069.1 | - | 11 - 12 | 1088 | 71 | |
AK300176.1 | - | 10 - 11 | 1037 | 71 | |
AK300842.1 | - | 11 - 12 | 1045 | 71 | |
AK303114.1 | - | 10 - 11 | 1026 | 71 | |
AL831952.1 | - | 3 - 4 | 2030 | 71 | |
BC008356.1 | - | 11 - 12 | 1106 | 71 | |
BC008747.2 | - | 11 - 12 | 1075 | 71 | |
BC025679.1 | - | 11 - 12 | 1076 | 71 | |
BT006964.1 | - | 10 - 11 | 952 | 71 | |
D38255.1 | - | 11 - 12 | 1073 | 71 | |
X80198.1 | - | 11 - 12 | 1061 | 71 |