The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001165937.1 | NP_001159409.1 | 5 - 6 | 612 | 65 |
NM_006804.3 | NP_006795.3 | 5 - 6 | 625 | 65 | |
XM_006721646.2 | XP_006721709.1 | 5 - 6 | 594 | 65 | |
XM_011524210.1 | XP_011522512.1 | 5 - 6 | 594 | 65 | |
XM_011524211.2 | XP_011522513.1 | 5 - 6 | 597 | 65 | |
XM_011524213.1 | XP_011522515.1 | 5 - 6 | 594 | 65 | |
XM_017024041.1 | XP_016879530.1 | 5 - 6 | 597 | 65 | |
GenBank mRNA | AK225069.1 | - | 5 - 6 | 558 | 65 |
AK300842.1 | - | 5 - 6 | 502 | 65 | |
BC008356.1 | - | 5 - 6 | 576 | 65 | |
BC008747.2 | - | 5 - 6 | 545 | 65 | |
BC025679.1 | - | 5 - 6 | 546 | 65 | |
BT006964.1 | - | 4 - 5 | 422 | 65 | |
D38255.1 | - | 5 - 6 | 543 | 65 | |
X80198.1 | - | 5 - 6 | 531 | 65 |