The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001030002.2 | NP_001025173.1 | 6 - 7 | 785 | 64 |
NM_001242442.1 | NP_001229371.1 | 6 - 7 | 869 | 64 | |
NM_001242443.1 | NP_001229372.1 | 6 - 7 | 930 | 64 | |
NM_005310.3 | NP_005301.2 | 6 - 7 | 940 | 64 | |
XM_017024536.1 | XP_016880025.1 | 5 - 6 | 2059 | 64 | |
XM_017024537.1 | XP_016880026.1 | 6 - 7 | 907 | 64 | |
XM_017024538.1 | XP_016880027.1 | 5 - 6 | 1107 | 64 | |
GenBank mRNA | AB008789.1 | - | 5 - 6 | 716 | 64 |
AB008790.1 | - | 5 - 6 | 716 | 64 | |
AK027729.1 | - | 6 - 7 | 785 | 64 | |
AK125544.1 | - | 6 - 7 | 64 | ||
AK222849.1 | - | 6 - 7 | 888 | 64 | |
AK222870.1 | - | 6 - 7 | 887 | 64 | |
AK290115.1 | - | 6 - 7 | 930 | 64 | |
AK294620.1 | - | 5 - 6 | 957 | 64 | |
AK314368.1 | - | 6 - 7 | 922 | 64 | |
BC006535.2 | - | 6 - 7 | 771 | 64 | |
BT006686.1 | - | 5 - 6 | 665 | 64 | |
D43772.1 | - | 6 - 7 | 884 | 64 | |
D87513.1 | - | 5 - 6 | 596 | 64 | |
KU178103.1 | - | 5 - 6 | 665 | 64 | |
KU178104.1 | - | 4 - 5 | 790 | 64 |