Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABLIM2
Entrez Gene ID
84448
Gene Name
actin binding LIM protein family member 2
Gene Aliases
-
Chromosome Location
Chr.4: 7965310 - 8158832 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130083.1 NP_001123555.1 4 - 5 596 95
NM_001130084.1 NP_001123556.1 4 - 5 596 95
NM_001130085.1 NP_001123557.1 4 - 5 596 95
NM_001130086.1 NP_001123558.1 4 - 5 596 95
NM_001130087.1 NP_001123559.1 4 - 5 596 95
NM_001130088.1 NP_001123560.1 4 - 5 596 95
NM_032432.4 NP_115808.3 4 - 5 596 95
XM_005248014.4 XP_005248071.1 4 - 5 567 95
XM_005248017.4 XP_005248074.1 4 - 5 567 95
XM_005248018.4 XP_005248075.1 4 - 5 566 95
XM_005248019.4 XP_005248076.1 4 - 5 566 95
XM_005248020.4 XP_005248077.1 4 - 5 566 95
XM_005248021.4 XP_005248078.1 4 - 5 565 95
XM_005248022.4 XP_005248079.1 4 - 5 566 95
XM_005248023.4 XP_005248080.1 4 - 5 566 95
XM_005248024.4 XP_005248081.1 4 - 5 565 95
XM_005248027.4 XP_005248084.1 4 - 5 565 95
XM_005248028.4 XP_005248085.1 4 - 5 562 95
XM_005248029.4 XP_005248086.1 4 - 5 564 95
XM_005248030.4 XP_005248087.1 4 - 5 564 95
XM_005248031.4 XP_005248088.1 4 - 5 566 95
XM_006713923.3 XP_006713986.1 4 - 5 567 95
XM_006713924.3 XP_006713987.1 4 - 5 566 95
XM_006713925.3 XP_006713988.1 4 - 5 567 95
XM_006713927.3 XP_006713990.1 4 - 5 565 95
XM_011513586.2 XP_011511888.1 4 - 5 571 95
XM_017008706.1 XP_016864195.1 4 - 5 568 95
XM_017008707.1 XP_016864196.1 4 - 5 577 95
XM_017008708.1 XP_016864197.1 4 - 5 566 95
XM_017008709.1 XP_016864198.1 4 - 5 567 95
XM_017008710.1 XP_016864199.1 4 - 5 566 95
XM_017008711.1 XP_016864200.1 4 - 5 566 95
XM_017008712.1 XP_016864201.1 4 - 5 566 95
XM_017008713.1 XP_016864202.1 4 - 5 565 95
XM_017008714.1 XP_016864203.1 4 - 5 577 95
XM_017008715.1 XP_016864204.1 4 - 5 566 95
XM_017008716.1 XP_016864205.1 4 - 5 565 95
XM_017008717.1 XP_016864206.1 4 - 5 565 95
XM_017008718.1 XP_016864207.1 4 - 5 577 95
XM_017008719.1 XP_016864208.1 4 - 5 565 95
XM_017008720.1 XP_016864209.1 4 - 5 564 95
XM_017008721.1 XP_016864210.1 4 - 5 564 95
XM_017008722.1 XP_016864211.1 4 - 5 565 95
XM_017008723.1 XP_016864212.1 4 - 5 577 95
XM_017008724.1 XP_016864213.1 4 - 5 577 95
XM_017008725.1 XP_016864214.1 4 - 5 577 95
GenBank mRNA AB058711.1 - 3 - 4 355 95
AJ748600.1 - 4 - 5 518 95
AJ748601.1 - 4 - 5 518 95
AK094754.1 - 4 - 5 575 95
AK301060.1 - 4 - 5 559 95
BC067214.1 - 4 - 5 608 95
BC122567.1 - 4 - 5 547 95
DQ413176.1 - 4 - 5 473 95
DQ413177.1 - 3 - 4 453 95

View Full Product Details