The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052920.1 | NP_443152.1 | 4 - 5 | 934 | 112 |
XM_006711929.3 | XP_006711992.1 | 3 - 4 | 2030 | 112 | |
XM_017003264.1 | XP_016858753.1 | 3 - 4 | 834 | 112 | |
GenBank mRNA | AL834515.1 | - | 2 - 3 | 185 | 112 |
BC069271.1 | - | 3 - 4 | 508 | 112 | |
DR002067.1 | - | 3 - 4 | 573 | 112 |