Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASB10
Entrez Gene ID
136371
Gene Name
ankyrin repeat and SOCS box containing 10
Gene Aliases
GLC1F
Chromosome Location
Chr.7: 151175698 - 151187832 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142459.1 NP_001135931.2 3 - 4 1361 107
NM_080871.3 NP_543147.2 3 - 4 1165 107
GenBank mRNA AK055536.1 - 3 - 4 1071 107
AK125565.1 - 3 - 4 1165 107
AK315114.1 - 3 - 4 1191 107
BC126351.1 - 3 - 4 1125 107

View Full Product Details