The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142459.1 | NP_001135931.2 | 4 - 5 | 1478 | 62 |
NM_080871.3 | NP_543147.2 | 4 - 5 | 1282 | 62 | |
GenBank mRNA | AK055536.1 | - | 4 - 5 | 1188 | 62 |
AK125565.1 | - | 4 - 5 | 1282 | 62 | |
AK315114.1 | - | 4 - 5 | 1308 | 62 | |
BC126351.1 | - | 4 - 5 | 1242 | 62 |