Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PNOC
Entrez Gene ID
5368
Gene Name
prepronociceptin
Gene Aliases
N/OFQ, NOP, OFQ, PPNOC, ppN/OFQ
Chromosome Location
Chr.8: 28316979 - 28343355 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011544559.2 XP_011542861.1 1 - 2 76 74

View Full Product Details