The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286072.1 | NP_001273001.1 | 2 - 3 | 639 | 71 |
XM_005270869.4 | XP_005270926.1 | 2 - 3 | 449 | 71 | |
XM_005270870.4 | XP_005270927.1 | 2 - 3 | 465 | 71 | |
XM_017001318.1 | XP_016856807.1 | 2 - 3 | 449 | 71 | |
GenBank mRNA | AB046851.1 | - | 2 - 3 | 253 | 71 |
AK308874.1 | - | 1 - 2 | 134 | 71 | |
BX647787.1 | - | 2 - 3 | 231 | 71 |