The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007553.2 | NP_001007554.1 | 11 - 12 | 1700 | 108 |
NM_001130523.2 | NP_001123995.1 | 11 - 12 | 1754 | 108 | |
NM_001242891.1 | NP_001229820.1 | 12 - 13 | 1847 | 108 | |
NM_001242892.1 | NP_001229821.1 | 10 - 11 | 1313 | 108 | |
NM_001242893.1 | NP_001229822.1 | 9 - 10 | 1220 | 108 | |
NM_007158.5 | NP_009089.4 | 10 - 11 | 1607 | 108 | |
GenBank mRNA | AB020692.1 | - | 11 - 12 | 1614 | 108 |
AF070542.1 | - | 9 - 10 | 1204 | 108 | |
AK022516.1 | - | 9 - 10 | 1122 | 108 | |
AK022528.1 | - | 9 - 10 | 1122 | 108 | |
AK290146.1 | - | 10 - 11 | 1245 | 108 | |
AY049788.1 | - | 8 - 9 | 1109 | 108 | |
BC032446.1 | - | 10 - 11 | 1542 | 108 | |
BX648561.1 | - | 10 - 11 | 1444 | 108 | |
BX648780.1 | - | 10 - 11 | 1541 | 108 | |
CR749378.1 | - | 11 - 12 | 1704 | 108 | |
CR749431.1 | - | 11 - 12 | 1686 | 108 |