Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CSDE1
Entrez Gene ID
7812
Gene Name
cold shock domain containing E1
Gene Aliases
D1S155E, UNR
Chromosome Location
Chr.1: 114716913 - 114758050 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007553.2 NP_001007554.1 13 - 14 1977 79
NM_001130523.2 NP_001123995.1 13 - 14 2031 79
NM_001242891.1 NP_001229820.1 14 - 15 2124 79
NM_001242892.1 NP_001229821.1 12 - 13 1590 79
NM_001242893.1 NP_001229822.1 11 - 12 1497 79
NM_007158.5 NP_009089.4 12 - 13 1884 79
GenBank mRNA AB020692.1 - 13 - 14 1891 79
AF077054.1 - 12 - 13 1800 79
AK022516.1 - 11 - 12 1399 79
AK022528.1 - 11 - 12 1399 79
AK130392.1 - 1 - 2 85 79
AK290146.1 - 12 - 13 1522 79
AL050150.1 - 2 - 3 177 79
AY049788.1 - 10 - 11 1386 79
BC032446.1 - 12 - 13 1819 79
BX648561.1 - 12 - 13 1721 79
CR749378.1 - 13 - 14 1981 79
CR749431.1 - 13 - 14 1963 79

View Full Product Details