The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007553.2 | NP_001007554.1 | 8 - 9 | 1223 | 76 |
NM_001130523.2 | NP_001123995.1 | 8 - 9 | 1277 | 76 | |
NM_001242891.1 | NP_001229820.1 | 9 - 10 | 1370 | 76 | |
NM_001242892.1 | NP_001229821.1 | 7 - 8 | 836 | 76 | |
NM_001242893.1 | NP_001229822.1 | 6 - 7 | 743 | 76 | |
NM_007158.5 | NP_009089.4 | 7 - 8 | 1130 | 76 | |
GenBank mRNA | AB020692.1 | - | 8 - 9 | 1137 | 76 |
AF070542.1 | - | 6 - 7 | 727 | 76 | |
AK022516.1 | - | 6 - 7 | 645 | 76 | |
AK022528.1 | - | 6 - 7 | 645 | 76 | |
AK290146.1 | - | 7 - 8 | 768 | 76 | |
AY049788.1 | - | 5 - 6 | 632 | 76 | |
BC032446.1 | - | 7 - 8 | 1065 | 76 | |
BX648561.1 | - | 7 - 8 | 967 | 76 | |
CR749378.1 | - | 8 - 9 | 1227 | 76 | |
CR749431.1 | - | 8 - 9 | 1209 | 76 |