The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242673.1 | NP_001229602.1 | 14 - 15 | 2043 | 73 |
NM_001242674.1 | NP_001229603.1 | 15 - 16 | 1413 | 73 | |
NM_001242675.1 | NP_001229604.1 | 14 - 15 | 1767 | 73 | |
NM_001242676.1 | NP_001229605.1 | 14 - 15 | 1756 | 73 | |
NM_006621.5 | NP_006612.2 | 14 - 15 | 1672 | 73 | |
XM_011540535.1 | XP_011538837.1 | 14 - 15 | 1695 | 73 | |
GenBank mRNA | AF035319.1 | - | 6 - 7 | 447 | 73 |
AF315687.1 | - | 14 - 15 | 1751 | 73 | |
AK131563.1 | - | 10 - 11 | 2251 | 73 | |
AK303690.1 | - | 14 - 15 | 1319 | 73 | |
AK316110.1 | - | 14 - 15 | 1571 | 73 | |
AL049954.1 | - | 14 - 15 | 1586 | 73 | |
BC003631.2 | - | 5 - 6 | 392 | 73 | |
BC007576.1 | - | 14 - 15 | 1600 | 73 | |
BC010681.1 | - | 14 - 15 | 1659 | 73 | |
BC016942.1 | - | 14 - 15 | 1630 | 73 | |
BC065254.1 | - | 14 - 15 | 1663 | 73 | |
BC095476.1 | - | 14 - 15 | 1615 | 73 | |
BC110896.1 | - | 14 - 15 | 1590 | 73 | |
BC111563.1 | - | 14 - 15 | 1309 | 73 | |
U82761.1 | - | 14 - 15 | 1340 | 73 |