Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BRPF3
Entrez Gene ID
27154
Gene Name
bromodomain and PHD finger containing 3
Gene Aliases
-
Chromosome Location
Chr.6: 36196773 - 36232790 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015695.2 NP_056510.2 8 - 9 3214 96
XM_005249010.2 XP_005249067.1 8 - 9 3096 96
XM_005249011.3 XP_005249068.1 8 - 9 3220 96
XM_011514489.1 XP_011512791.1 8 - 9 3118 96
XM_011514490.1 XP_011512792.1 7 - 8 3016 96
XM_011514491.1 XP_011512793.1 7 - 8 3016 96
XM_017010742.1 XP_016866231.1 7 - 8 3016 96
XM_017010743.1 XP_016866232.1 5 - 6 1332 96
GenBank mRNA AB033112.1 - 8 - 9 3214 96
AK025134.1 - 1 - 2 274 96
BC033652.2 - 7 - 8 2933 96

View Full Product Details