The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000063.5 | NP_000054.2 | 13 - 14 | 2009 | 74 |
NM_001145903.2 | NP_001139375.1 | 11 - 12 | 1613 | 74 | |
NM_001178063.2 | NP_001171534.1 | 9 - 10 | 1159 | 74 | |
NM_001282457.1 | NP_001269386.1 | 9 - 10 | 1216 | 74 | |
NM_001282458.1 | NP_001269387.1 | 13 - 14 | 2037 | 74 | |
GenBank mRNA | AK096258.1 | - | 10 - 11 | 1314 | 74 |
AK222537.1 | - | 13 - 14 | 1761 | 74 | |
AK222783.1 | - | 13 - 14 | 1733 | 74 | |
AK298311.1 | - | 9 - 10 | 1159 | 74 | |
AK298808.1 | - | 9 - 10 | 1216 | 74 | |
AK300892.1 | - | 11 - 12 | 1373 | 74 | |
AK300930.1 | - | 13 - 14 | 1775 | 74 | |
AK307930.1 | - | 11 - 12 | 1367 | 74 | |
AK312581.1 | - | 13 - 14 | 1843 | 74 | |
BC043484.1 | - | 13 - 14 | 1744 | 74 | |
BX537504.1 | - | 7 - 8 | 1746 | 74 | |
X04481.1 | - | 13 - 14 | 1769 | 74 |