The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000063.5 | NP_000054.2 | 6 - 7 | 1120 | 93 |
NM_001145903.2 | NP_001139375.1 | 4 - 5 | 724 | 93 | |
NM_001282458.1 | NP_001269387.1 | 6 - 7 | 1148 | 93 | |
GenBank mRNA | AK222537.1 | - | 6 - 7 | 872 | 93 |
AK222783.1 | - | 6 - 7 | 844 | 93 | |
AK300892.1 | - | 4 - 5 | 484 | 93 | |
AK300930.1 | - | 6 - 7 | 93 | ||
AK307930.1 | - | 4 - 5 | 480 | 93 | |
AK312581.1 | - | 6 - 7 | 954 | 93 | |
BC043484.1 | - | 6 - 7 | 855 | 93 | |
X04481.1 | - | 6 - 7 | 880 | 93 |