The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321995.1 | NP_001308924.1 | 2 - 3 | 197 | 86 |
NM_001321996.1 | NP_001308925.1 | 2 - 3 | 376 | 86 | |
NM_001321997.1 | NP_001308926.1 | 2 - 3 | 197 | 86 | |
NM_031902.4 | NP_114108.1 | 2 - 3 | 376 | 86 | |
NR_136152.1 | 2 - 3 | 376 | 86 | ||
NR_136153.1 | 2 - 3 | 376 | 86 | ||
GenBank mRNA | AB049940.1 | - | 2 - 3 | 257 | 86 |
AK058160.1 | - | 2 - 3 | 150 | 86 | |
AK295812.1 | - | 2 - 3 | 162 | 86 | |
BC014172.2 | - | 2 - 3 | 349 | 86 | |
BG771419.1 | - | 2 - 3 | 363 | 86 |