The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321995.1 | NP_001308924.1 | 8 - 9 | 870 | 153 |
NM_001321996.1 | NP_001308925.1 | 8 - 9 | 970 | 153 | |
NM_001321997.1 | NP_001308926.1 | 8 - 9 | 791 | 153 | |
NM_031902.4 | NP_114108.1 | 8 - 9 | 1049 | 153 | |
NR_136152.1 | 7 - 8 | 879 | 153 | ||
NR_136153.1 | 7 - 8 | 958 | 153 | ||
GenBank mRNA | AB049940.1 | - | 8 - 9 | 930 | 153 |
AK058160.1 | - | 7 - 8 | 732 | 153 | |
BC000219.2 | - | 4 - 5 | 364 | 153 | |
BC014172.2 | - | 8 - 9 | 1022 | 153 |