The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321995.1 | NP_001308924.1 | 10 - 11 | 993 | 158 |
NM_001321996.1 | NP_001308925.1 | 10 - 11 | 1093 | 158 | |
NM_001321997.1 | NP_001308926.1 | 10 - 11 | 914 | 158 | |
NM_031902.4 | NP_114108.1 | 10 - 11 | 1172 | 158 | |
NR_136152.1 | 9 - 10 | 1002 | 158 | ||
NR_136153.1 | 9 - 10 | 1081 | 158 | ||
GenBank mRNA | AB049940.1 | - | 10 - 11 | 1053 | 158 |
AK058160.1 | - | 9 - 10 | 855 | 158 | |
BC000219.2 | - | 6 - 7 | 487 | 158 | |
BC014172.2 | - | 10 - 11 | 1145 | 158 |