Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GHDC
Entrez Gene ID
84514
Gene Name
GH3 domain containing
Gene Aliases
D11LGP1, LGP1
Chromosome Location
Chr.17: 42189087 - 42194532 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032484.4 NP_115873.1 8 - 9 1520 78
GenBank mRNA AF316997.1 - 6 - 7 1296 78
AK074700.1 - 7 - 8 1724 78
AK122777.1 - 9 - 10 1288 78
AK123075.1 - - 1538 78
AK298933.1 - 8 - 9 1281 78
BC022784.1 - 8 - 9 1455 78

View Full Product Details