The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320802.1 | NP_001307731.1 | 1 - 2 | 321 | 63 |
GenBank mRNA | AK075212.1 | - | 1 - 2 | 78 | 63 |
BC017087.1 | - | 1 - 2 | 92 | 63 | |
BC071833.1 | - | 1 - 2 | 71 | 63 |