The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206480.2 | NP_001193409.1 | 13 - 14 | 1621 | 67 |
NM_001206482.1 | NP_001193411.1 | 13 - 14 | 1791 | 67 | |
NM_014800.10 | NP_055615.8 | 13 - 14 | 1432 | 67 | |
XM_005249919.2 | XP_005249976.1 | 14 - 15 | 1985 | 67 | |
XM_006715805.1 | XP_006715868.1 | 13 - 14 | 1277 | 67 | |
XM_011515654.1 | XP_011513956.1 | 14 - 15 | 2670 | 67 | |
XM_017012838.1 | XP_016868327.1 | 14 - 15 | 2855 | 67 | |
XM_017012839.1 | XP_016868328.1 | 13 - 14 | 1219 | 67 | |
GenBank mRNA | AF398885.1 | - | 12 - 13 | 1083 | 67 |
AK291784.1 | - | 13 - 14 | 1791 | 67 | |
AK296253.1 | - | 10 - 11 | 952 | 67 | |
AK314349.1 | - | 13 - 14 | 1221 | 67 | |
AL136787.1 | - | 13 - 14 | 1333 | 67 | |
BC114341.2 | - | 13 - 14 | 1170 | 67 |