Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CPS1
Entrez Gene ID
1373
Gene Name
carbamoyl-phosphate synthase 1
Gene Aliases
CPSASE1, PHN
Chromosome Location
Chr.2: 210477682 - 210679107 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122633.2 NP_001116105.1 22 - 23 2790 127
NM_001122634.3 NP_001116106.1 11 - 12 1846 127
NM_001875.4 NP_001866.2 21 - 22 2825 127
XM_011510640.1 XP_011508942.1 23 - 24 3024 127
XM_011510641.2 XP_011508943.1 23 - 24 3168 127
XM_011510642.2 XP_011508944.1 22 - 23 3040 127
XM_011510643.2 XP_011508945.1 23 - 24 2905 127
XM_011510644.2 XP_011508946.1 22 - 23 2946 127
GenBank mRNA AB180934.1 - 21 - 23 127
AB180935.1 - 21 - 23 127
AB180936.1 - 21 - 23 127
AB180937.1 - 21 - 23 127
AB180938.1 - 21 - 23 127
AB208800.1 - 22 - 24 127
AF154830.1 - 21 - 23 127
AK289749.1 - 21 - 23 127
AK302778.1 - 11 - 13 127
AY317138.1 - 11 - 12 1832 127
BC140943.1 - 21 - 22 2792 127
D90282.1 - 21 - 22 2808 127
Y15793.1 - 21 - 22 2690 127

View Full Product Details