Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CPS1
Entrez Gene ID
1373
Gene Name
carbamoyl-phosphate synthase 1
Gene Aliases
CPSASE1, PHN
Chromosome Location
Chr.2: 210477682 - 210679107 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122633.2 NP_001116105.1 24 - 25 2997 101
NM_001122634.3 NP_001116106.1 13 - 14 2053 101
NM_001875.4 NP_001866.2 23 - 24 3032 101
XM_011510640.1 XP_011508942.1 25 - 26 3231 101
XM_011510641.2 XP_011508943.1 25 - 26 3375 101
XM_011510642.2 XP_011508944.1 24 - 25 3247 101
XM_011510643.2 XP_011508945.1 25 - 26 3112 101
XM_011510644.2 XP_011508946.1 24 - 25 3153 101
GenBank mRNA AB180934.1 - 24 - 25 2897 101
AB180935.1 - 24 - 25 2897 101
AB180936.1 - 24 - 25 2897 101
AB180938.1 - 24 - 25 2897 101
AB208800.1 - 25 - 26 2973 101
AF154830.1 - 24 - 25 3020 101
AK289749.1 - 24 - 25 3032 101
AK302778.1 - 14 - 15 2042 101
AY317138.1 - 13 - 14 2039 101
BC140943.1 - 23 - 24 2999 101
BX640601.1 - 5 - 6 517 101
D90282.1 - 23 - 24 3015 101
Y15793.1 - 23 - 24 2897 101

View Full Product Details