The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030623.3 | NP_085126.2 | 8 - 9 | 4672 | 97 |
XM_006712778.3 | XP_006712841.1 | 9 - 10 | 5104 | 97 | |
GenBank mRNA | AB051465.1 | - | 2 - 3 | 3520 | 97 |
AK090876.1 | - | 2 - 3 | 1747 | 97 | |
CR749494.1 | - | 8 - 9 | 4654 | 97 |