The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001307976.1 | NP_001294905.1 | 11 - 12 | 2578 | 84 |
NM_152519.3 | NP_689732.2 | 12 - 13 | 2704 | 84 | |
XM_005246328.4 | XP_005246385.1 | 12 - 13 | 2986 | 84 | |
XM_005246329.4 | XP_005246386.1 | 12 - 13 | 2692 | 84 | |
XM_005246330.4 | XP_005246387.1 | 12 - 13 | 2573 | 84 | |
XM_005246332.4 | XP_005246389.1 | 11 - 12 | 2859 | 84 | |
XM_011510706.2 | XP_011509008.1 | 12 - 13 | 2611 | 84 | |
XM_011510707.2 | XP_011509009.1 | 12 - 13 | 2794 | 84 | |
XM_011510710.2 | XP_011509012.1 | 11 - 12 | 1443 | 84 | |
XM_017003430.1 | XP_016858919.1 | 12 - 13 | 2730 | 84 | |
XM_017003431.1 | XP_016858920.1 | 8 - 9 | 4812 | 84 | |
GenBank mRNA | AK074441.1 | - | 11 - 12 | 1474 | 84 |
AL133053.1 | - | 6 - 7 | 696 | 84 | |
BC090854.1 | - | 11 - 12 | 2041 | 84 | |
BC126155.1 | - | 11 - 12 | 2484 | 84 | |
BC143914.1 | - | 11 - 12 | 2484 | 84 |