The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031701.2 | NP_001026871.1 | 7 - 8 | 850 | 83 |
XM_011538476.2 | XP_011536778.1 | 4 - 5 | 607 | 83 | |
XM_017019454.1 | XP_016874943.1 | 7 - 8 | 891 | 83 | |
GenBank mRNA | AB032773.1 | - | 7 - 8 | 660 | 83 |
BC052293.1 | - | 7 - 8 | 850 | 83 |