Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
APAF1
Entrez Gene ID
317
Gene Name
apoptotic peptidase activating factor 1
Gene Aliases
APAF-1, CED4
Chromosome Location
Chr.12: 98645141 - 98735433 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160.2 NP_001151.1 14 - 15 2589 105
NM_013229.2 NP_037361.1 14 - 15 2589 105
NM_181861.1 NP_863651.1 14 - 15 2622 105
NM_181868.1 NP_863658.1 14 - 15 2622 105
GenBank mRNA AB007873.2 - 13 - 14 2012 105
AF013263.1 - 14 - 15 2589 105
AF134397.1 - 13 - 14 2012 105
AF149794.1 - 13 - 14 2045 105
AJ243003.1 - 13 - 14 2045 105
AJ243004.1 - 13 - 14 2045 105
AJ243005.1 - 13 - 14 2045 105
AJ243006.1 - 13 - 14 2045 105
AJ243007.1 - 13 - 14 2045 105
AJ243008.1 - 13 - 14 2045 105
AJ243009.1 - 13 - 14 2045 105
AJ243010.1 - 13 - 14 2045 105
AJ243011.1 - 13 - 14 2045 105
AJ243048.1 - 13 - 14 2045 105
AJ243107.1 - 13 - 14 2069 105
BC136531.1 - 14 - 15 2193 105
BC136532.1 - 14 - 15 2193 105
BC150255.1 - 13 - 14 2041 105
EF560718.1 - 13 - 14 2050 105

View Full Product Details