The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143819.2 | NP_001137291.1 | 10 - 11 | 1251 | 64 |
NM_001301214.1 | NP_001288143.1 | 8 - 9 | 833 | 64 | |
NM_017901.5 | NP_060371.2 | 9 - 10 | 1158 | 64 | |
XM_005253906.4 | XP_005253963.1 | 8 - 9 | 1672 | 64 | |
XM_011538490.2 | XP_011536792.1 | 10 - 11 | 1217 | 64 | |
XM_011538492.2 | XP_011536794.1 | 9 - 10 | 1064 | 64 | |
XM_011538493.2 | XP_011536795.1 | 8 - 9 | 908 | 64 | |
XM_017019480.1 | XP_016874969.1 | 10 - 11 | 1177 | 64 | |
XM_017019481.1 | XP_016874970.1 | 10 - 11 | 1263 | 64 | |
XM_017019482.1 | XP_016874971.1 | 10 - 11 | 1178 | 64 | |
GenBank mRNA | AB032995.2 | - | 9 - 10 | 1125 | 64 |
AK057414.1 | - | 10 - 11 | 1199 | 64 | |
AK093490.1 | - | 9 - 10 | 1158 | 64 | |
AK295022.1 | - | 8 - 9 | 1025 | 64 | |
AK296268.1 | - | 8 - 9 | 833 | 64 | |
AY083666.1 | - | 9 - 10 | 1131 | 64 | |
BC136795.1 | - | 9 - 10 | 1115 | 64 | |
BC136796.1 | - | 9 - 10 | 1115 | 64 | |
BC142664.1 | - | 9 - 10 | 1110 | 64 | |
BC150203.1 | - | 9 - 10 | 1081 | 64 | |
BC152423.1 | - | 9 - 10 | 1125 | 64 |