The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143819.2 | NP_001137291.1 | 16 - 17 | 1744 | 97 |
NM_001301214.1 | NP_001288143.1 | 14 - 15 | 1326 | 97 | |
NM_017901.5 | NP_060371.2 | 15 - 16 | 1651 | 97 | |
XM_005253906.4 | XP_005253963.1 | 14 - 15 | 2165 | 97 | |
XM_011538490.2 | XP_011536792.1 | 16 - 17 | 1710 | 97 | |
XM_011538492.2 | XP_011536794.1 | 15 - 16 | 1557 | 97 | |
XM_011538493.2 | XP_011536795.1 | 14 - 15 | 1401 | 97 | |
XM_017019480.1 | XP_016874969.1 | 16 - 17 | 1670 | 97 | |
XM_017019481.1 | XP_016874970.1 | 16 - 17 | 1756 | 97 | |
XM_017019482.1 | XP_016874971.1 | 16 - 17 | 1671 | 97 | |
GenBank mRNA | AB032995.2 | - | 15 - 16 | 1618 | 97 |
AK057414.1 | - | 16 - 17 | 1692 | 97 | |
AK075071.1 | - | 5 - 6 | 311 | 97 | |
AK093490.1 | - | 15 - 16 | 1651 | 97 | |
AK295022.1 | - | 14 - 15 | 1518 | 97 | |
AK296268.1 | - | 14 - 15 | 1326 | 97 | |
AY083666.1 | - | 15 - 16 | 1624 | 97 | |
BC136795.1 | - | 15 - 16 | 1608 | 97 | |
BC136796.1 | - | 15 - 16 | 1608 | 97 | |
BC142664.1 | - | 15 - 16 | 1603 | 97 | |
BC150203.1 | - | 15 - 16 | 1574 | 97 | |
BC152423.1 | - | 15 - 16 | 1618 | 97 |