The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143819.2 | NP_001137291.1 | 22 - 23 | 2201 | 67 |
NM_001301214.1 | NP_001288143.1 | 20 - 21 | 1783 | 67 | |
NM_017901.5 | NP_060371.2 | 21 - 22 | 2108 | 67 | |
XM_005253906.4 | XP_005253963.1 | 20 - 21 | 2622 | 67 | |
XM_011538490.2 | XP_011536792.1 | 22 - 23 | 2167 | 67 | |
XM_011538492.2 | XP_011536794.1 | 21 - 22 | 2014 | 67 | |
XM_011538493.2 | XP_011536795.1 | 20 - 21 | 1858 | 67 | |
XM_017019480.1 | XP_016874969.1 | 21 - 22 | 2073 | 67 | |
XM_017019481.1 | XP_016874970.1 | 22 - 23 | 2213 | 67 | |
GenBank mRNA | AB032995.2 | - | 21 - 22 | 2075 | 67 |
AK057414.1 | - | 22 - 23 | 2149 | 67 | |
AK075071.1 | - | 10 - 11 | 1291 | 67 | |
AK093490.1 | - | 21 - 22 | 2108 | 67 | |
AK296268.1 | - | 20 - 21 | 1783 | 67 | |
AY083666.1 | - | 21 - 22 | 2081 | 67 | |
BC136795.1 | - | 21 - 22 | 2065 | 67 | |
BC136796.1 | - | 21 - 22 | 2065 | 67 | |
BC142664.1 | - | 20 - 21 | 2583 | 67 | |
BC150203.1 | - | 21 - 22 | 2031 | 67 | |
BC152423.1 | - | 21 - 22 | 2075 | 67 |