The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001131028.1 | NP_001124500.1 | 6 - 7 | 882 | 84 |
NM_031482.4 | NP_113670.1 | 5 - 6 | 747 | 84 | |
XM_005248610.4 | XP_005248667.1 | 5 - 6 | 2017 | 84 | |
XM_005248611.4 | XP_005248668.1 | 5 - 6 | 591 | 84 | |
XM_005248612.3 | XP_005248669.1 | 4 - 5 | 643 | 84 | |
XM_011543660.2 | XP_011541962.1 | 4 - 5 | 447 | 84 | |
XM_011543661.2 | XP_011541963.1 | 3 - 4 | 830 | 84 | |
XM_017009944.1 | XP_016865433.1 | 5 - 6 | 778 | 84 | |
GenBank mRNA | AK225969.1 | - | 4 - 5 | 639 | 84 |
AK315752.1 | - | 5 - 6 | 747 | 84 | |
AL136912.1 | - | 5 - 6 | 615 | 84 | |
BC029268.1 | - | 5 - 6 | 717 | 84 |