The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001247997.1 | NP_001234926.1 | 24 - 25 | 4120 | 113 |
NM_002956.2 | NP_002947.1 | 23 - 24 | 4087 | 113 | |
NM_198240.1 | NP_937883.1 | 22 - 23 | 3982 | 113 | |
XM_017019786.1 | XP_016875275.1 | 23 - 24 | 4251 | 113 | |
XM_017019790.1 | XP_016875279.1 | 22 - 23 | 3972 | 113 | |
GenBank mRNA | BC114213.1 | - | 21 - 22 | 3897 | 113 |
BC117209.1 | - | 23 - 24 | 4007 | 113 | |
BC126305.1 | - | 23 - 24 | 4007 | 113 | |
M97501.1 | - | 22 - 23 | 3982 | 113 | |
X64838.1 | - | 23 - 24 | 4064 | 113 |