Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLIP1
Entrez Gene ID
6249
Gene Name
CAP-Gly domain containing linker protein 1
Gene Aliases
CLIP, CLIP-170, CLIP170, CYLN1, RSN
Chromosome Location
Chr.12: 122271432 - 122422956 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001247997.1 NP_001234926.1 7 - 8 1460 85
NM_002956.2 NP_002947.1 7 - 8 1460 85
NM_198240.1 NP_937883.1 7 - 8 1460 85
XM_005253593.2 XP_005253650.1 7 - 8 1523 85
XM_006719552.3 XP_006719615.1 7 - 8 1549 85
XM_017019786.1 XP_016875275.1 7 - 8 1624 85
XM_017019787.1 XP_016875276.1 7 - 8 1523 85
XM_017019788.1 XP_016875277.1 7 - 8 1549 85
XM_017019789.1 XP_016875278.1 7 - 8 1708 85
XM_017019790.1 XP_016875279.1 7 - 8 1450 85
XM_017019791.1 XP_016875280.1 7 - 8 1549 85
XM_017019792.1 XP_016875281.1 7 - 8 1549 85
GenBank mRNA AB209115.1 - 4 - 5 558 85
BC047895.1 - 6 - 7 1268 85
BC062543.1 - 6 - 7 1416 85
BC114213.1 - 6 - 7 1375 85
BC117209.1 - 6 - 7 1347 85
BC126305.1 - 6 - 7 1347 85
M97501.1 - 7 - 8 1460 85
X64838.1 - 7 - 8 1437 85

View Full Product Details