The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198240.1 | NP_937883.1 | 8 - 9 | 1525 | 98 |
XM_017019788.1 | XP_016875277.1 | 8 - 9 | 1614 | 98 | |
XM_017019789.1 | XP_016875278.1 | 8 - 9 | 1773 | 98 | |
XM_017019790.1 | XP_016875279.1 | 8 - 9 | 1515 | 98 | |
XM_017019792.1 | XP_016875281.1 | 8 - 9 | 1614 | 98 | |
GenBank mRNA | AY563000.1 | - | 2 - 3 | 95 | 98 |
BC114213.1 | - | 7 - 8 | 1440 | 98 | |
M97501.1 | - | 8 - 9 | 1525 | 98 |