Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLIP1
Entrez Gene ID
6249
Gene Name
CAP-Gly domain containing linker protein 1
Gene Aliases
CLIP, CLIP-170, CLIP170, CYLN1, RSN
Chromosome Location
Chr.12: 122271432 - 122422956 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001247997.1 NP_001234926.1 4 - 5 933 70
NM_002956.2 NP_002947.1 4 - 5 933 70
NM_198240.1 NP_937883.1 4 - 5 933 70
XM_005253593.2 XP_005253650.1 4 - 5 996 70
XM_006719552.3 XP_006719615.1 4 - 5 1022 70
XM_017019786.1 XP_016875275.1 4 - 5 1097 70
XM_017019787.1 XP_016875276.1 4 - 5 996 70
XM_017019788.1 XP_016875277.1 4 - 5 1022 70
XM_017019789.1 XP_016875278.1 4 - 5 1181 70
XM_017019790.1 XP_016875279.1 4 - 5 923 70
XM_017019791.1 XP_016875280.1 4 - 5 1022 70
XM_017019792.1 XP_016875281.1 4 - 5 1022 70
GenBank mRNA AB209115.1 - 1 - 2 31 70
BC047895.1 - 4 - 5 939 70
BC062543.1 - 3 - 4 889 70
BC114213.1 - 3 - 4 848 70
BC117209.1 - 3 - 4 820 70
BC126305.1 - 3 - 4 820 70
M97501.1 - 4 - 5 933 70
X64838.1 - 4 - 5 910 70

View Full Product Details