Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WFDC3
Entrez Gene ID
140686
Gene Name
WAP four-disulfide core domain 3
Gene Aliases
WAP14, dJ447F3.3
Chromosome Location
Chr.20: 45708860 - 45791908 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080614.1 NP_542181.1 3 - 4 290 67
XM_011528553.2 XP_011526855.1 3 - 4 300 67
GenBank mRNA AF488306.1 - 3 - 4 290 67
AL591713.1 - 2 - 3 187 67
BC026014.1 - 2 - 3 194 67
BC102013.1 - 2 - 3 196 67

View Full Product Details