The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003404.4 | NP_003395.1 | 4 - 5 | 803 | 125 |
NM_139323.3 | NP_647539.1 | 3 - 4 | 708 | 125 | |
XM_017028039.1 | XP_016883528.1 | 4 - 5 | 607 | 125 | |
GenBank mRNA | AB209759.1 | - | 2 - 3 | 4608 | 125 |
AB451260.1 | - | 2 - 3 | 417 | 125 | |
AB451386.1 | - | 2 - 3 | 417 | 125 | |
AF064847.1 | - | 4 - 5 | 727 | 125 | |
AK292717.1 | - | 4 - 5 | 696 | 125 | |
BC001359.2 | - | 3 - 4 | 598 | 125 | |
EU794580.1 | - | 4 - 5 | 699 | 125 | |
X57346.1 | - | 4 - 5 | 789 | 125 |