The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136566.2 | NP_001130038.2 | 3 - 4 | 373 | 118 |
XM_006723602.2 | XP_006723665.1 | 3 - 4 | 410 | 118 | |
XM_006723603.2 | XP_006723666.1 | 3 - 4 | 576 | 118 | |
XM_006723604.2 | XP_006723667.1 | 3 - 4 | 410 | 118 | |
XM_006723605.3 | XP_006723668.1 | 3 - 4 | 410 | 118 | |
XM_011529317.2 | XP_011527619.1 | 3 - 4 | 572 | 118 | |
GenBank mRNA | BC157891.1 | - | 3 - 4 | 118 | |
BC171911.1 | - | 3 - 4 | 118 | ||
HQ603827.1 | - | 2 - 3 | 118 |