The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135556.1 | NP_001129028.1 | 5 - 6 | 650 | 61 |
NM_004411.4 | NP_004402.1 | 5 - 6 | 701 | 61 | |
XM_011515861.1 | XP_011514163.1 | 5 - 6 | 548 | 61 | |
XM_011515862.1 | XP_011514164.1 | 5 - 6 | 497 | 61 | |
XM_011515863.2 | XP_011514165.1 | 5 - 6 | 655 | 61 | |
XM_017011804.1 | XP_016867293.1 | 5 - 6 | 1288 | 61 | |
XM_017011805.1 | XP_016867294.1 | 5 - 6 | 475 | 61 | |
XM_017011806.1 | XP_016867295.1 | 5 - 6 | 650 | 61 | |
XM_017011807.1 | XP_016867296.1 | 5 - 6 | 655 | 61 | |
GenBank mRNA | AF063228.1 | - | 5 - 6 | 611 | 61 |
AK091339.1 | - | 6 - 7 | 550 | 61 | |
AK309419.1 | - | 5 - 6 | 473 | 61 | |
AK313148.1 | - | 5 - 6 | 648 | 61 | |
BC032945.2 | - | 5 - 6 | 602 | 61 | |
BC037854.1 | - | 5 - 6 | 592 | 61 | |
BC053542.1 | - | 5 - 6 | 590 | 61 |