Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DYNC1I1
Entrez Gene ID
1780
Gene Name
dynein cytoplasmic 1 intermediate chain 1
Gene Aliases
DNCI1, DNCIC1
Chromosome Location
Chr.7: 95772506 - 96110322 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135556.1 NP_001129028.1 1 - 2 271 226
NM_001135557.1 NP_001129029.1 1 - 2 271 226
NM_001278421.1 NP_001265350.1 1 - 2 271 226
NM_001278422.1 NP_001265351.1 1 - 2 271 226
NM_004411.4 NP_004402.1 1 - 2 271 226
XM_011515863.2 XP_011514165.1 1 - 2 225 226
XM_017011806.1 XP_016867295.1 1 - 2 271 226
XM_017011807.1 XP_016867296.1 1 - 2 225 226
GenBank mRNA AF063228.1 - 1 - 2 181 223
AF123074.1 - 1 - 2 180 226
AK297427.1 - 1 - 2 183 226
AK313148.1 - 1 - 2 218 226
BC022540.1 - 1 - 2 180 226
BC032945.2 - 1 - 2 223 226
BC037854.1 - 1 - 2 213 226
BC053542.1 - 1 - 2 211 226
DA491512.1 - 1 - 2 271 226

View Full Product Details