Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAPGEF5
Entrez Gene ID
9771
Gene Name
Rap guanine nucleotide exchange factor 5
Gene Aliases
GFR, MR-GEF, REPAC
Chromosome Location
Chr.7: 22118290 - 22357133 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012294.3 NP_036426.3 13 - 14 1275 151
XM_005249914.2 XP_005249971.1 13 - 14 1494 151
XM_005249916.1 XP_005249973.1 13 - 14 1438 151
XM_011515652.2 XP_011513954.1 5 - 6 3248 151
XM_017012837.1 XP_016868326.1 11 - 12 1179 151
GenBank mRNA AA815190.1 - 2 - 3 151
AK295859.1 - 3 - 4 760 151
BC039203.1 - 3 - 4 586 151
BP349215.1 - 4 - 5 333 151
D87467.1 - 3 - 4 567 151
KU178490.1 - 3 - 4 512 151
KU178491.1 - 3 - 4 512 151

View Full Product Details