The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014432.1 | NP_001014432.1 | 2 - 3 | 342 | 120 |
NM_005163.2 | NP_005154.2 | 1 - 2 | 472 | 120 | |
XM_017021075.1 | XP_016876564.1 | 2 - 3 | 585 | 120 | |
XM_017021076.1 | XP_016876565.1 | 2 - 3 | 491 | 120 | |
XM_017021078.1 | XP_016876567.1 | 2 - 3 | 118 | 120 | |
GenBank mRNA | AK122894.1 | - | 1 - 2 | 1399 | 120 |
BC084538.1 | - | 2 - 3 | 336 | 120 | |
KF836747.1 | - | 2 - 3 | 236 | 120 | |
KF836748.1 | - | 2 - 3 | 211 | 120 | |
KF836749.1 | - | 2 - 3 | 142 | 120 | |
M63167.1 | - | 1 - 2 | 116 | 120 |