The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014431.1 | NP_001014431.1 | 8 - 9 | 1039 | 82 |
NM_001014432.1 | NP_001014432.1 | 9 - 10 | 1123 | 82 | |
NM_005163.2 | NP_005154.2 | 8 - 9 | 1253 | 82 | |
XM_005267401.1 | XP_005267458.1 | 8 - 9 | 1188 | 82 | |
XM_017021075.1 | XP_016876564.1 | 9 - 10 | 1366 | 82 | |
XM_017021076.1 | XP_016876565.1 | 9 - 10 | 1272 | 82 | |
XM_017021077.1 | XP_016876566.1 | 8 - 9 | 1188 | 82 | |
XM_017021078.1 | XP_016876567.1 | 9 - 10 | 899 | 82 | |
GenBank mRNA | AB451242.1 | - | 7 - 8 | 699 | 82 |
AB451367.1 | - | 7 - 8 | 699 | 82 | |
AK122894.1 | - | 8 - 9 | 2180 | 82 | |
AK299310.1 | - | 6 - 7 | 643 | 82 | |
AK314256.1 | - | 8 - 9 | 852 | 82 | |
BC000479.2 | - | 8 - 9 | 974 | 82 | |
BC084538.1 | - | 9 - 10 | 1117 | 82 | |
BX648205.1 | - | 7 - 8 | 1657 | 82 | |
KU177898.1 | - | 7 - 8 | 699 | 82 | |
M63167.1 | - | 8 - 9 | 897 | 82 |