The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015093.5 | NP_055908.1 | 3 - 4 | 333 | 95 |
XM_011535633.2 | XP_011533935.1 | 2 - 3 | 772 | 95 | |
XM_017010591.1 | XP_016866080.1 | 2 - 3 | 303 | 95 | |
GenBank mRNA | AK307650.1 | - | 3 - 4 | 320 | 95 |