The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130173.1 | NP_001123645.1 | 9 - 10 | 1406 | 151 |
NM_001161656.1 | NP_001155128.1 | 9 - 10 | 1397 | 151 | |
GenBank mRNA | AJ606319.1 | - | 9 - 10 | 1406 | 151 |
AY787455.1 | - | 10 - 11 | 1338 | 151 | |
AY787460.1 | - | 10 - 11 | 1320 | 151 | |
AY787461.1 | - | 9 - 10 | 1198 | 151 | |
AY787467.1 | - | 9 - 10 | 1207 | 151 |