The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173674.2 | NP_775945.1 | 4 - 5 | 691 | 74 |
XM_005266941.1 | XP_005266998.1 | 4 - 5 | 655 | 74 | |
XM_006715463.1 | XP_006715526.1 | 4 - 5 | 655 | 74 | |
XM_011535771.1 | XP_011534073.1 | 5 - 6 | 740 | 74 | |
XM_011535773.2 | XP_011534075.1 | 4 - 5 | 655 | 74 | |
XM_011535774.2 | XP_011534076.1 | 4 - 5 | 655 | 74 | |
XM_017010775.1 | XP_016866264.1 | 5 - 6 | 664 | 74 | |
GenBank mRNA | AK095973.1 | - | 4 - 5 | 561 | 74 |
BC035671.1 | - | 4 - 5 | 638 | 74 |