Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MICAL1
Entrez Gene ID
64780
Gene Name
microtubule associated monooxygenase, calponin and LIM domain containing 1
Gene Aliases
MICAL, MICAL-1, NICAL
Chromosome Location
Chr.6: 109444062 - 109465968 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159291.1 NP_001152763.1 15 - 16 2129 113
NM_001286613.1 NP_001273542.1 16 - 17 2423 113
NM_022765.3 NP_073602.3 16 - 17 2387 113
GenBank mRNA AB048948.1 - 16 - 17 2387 113
AK021999.1 - 15 - 16 2130 113
AK025392.1 - 16 - 17 2154 113
AK160384.1 - 13 - 14 3074 113
AK296284.1 - 16 - 17 2426 113
AL122098.1 - 8 - 9 813 113
BC009972.2 - 15 - 16 1884 113
BC042144.1 - 16 - 17 2283 113
BC052983.1 - 15 - 16 2100 113

View Full Product Details