The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152729.2 | NP_689942.2 | 6 - 7 | 620 | 156 |
XM_006715378.3 | XP_006715441.1 | 6 - 7 | 623 | 156 | |
GenBank mRNA | AK090918.1 | - | 3 - 4 | 1350 | 156 |
AK290408.1 | - | 6 - 7 | 570 | 156 | |
AK308337.1 | - | 6 - 7 | 603 | 156 | |
AY203935.1 | - | 3 - 4 | 230 | 156 | |
BC108699.1 | - | 6 - 7 | 577 | 156 | |
BC136832.1 | - | 6 - 7 | 579 | 156 | |
BC136841.1 | - | 6 - 7 | 579 | 156 | |
BM808222.1 | - | - | 609 | 156 |