The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152377.2 | NP_689590.1 | 7 - 8 | 1137 | 97 |
XM_017000307.1 | XP_016855796.1 | 6 - 7 | 947 | 97 | |
GenBank mRNA | AK124828.1 | - | 7 - 8 | 1122 | 97 |
BC027878.1 | - | 7 - 8 | 1121 | 97 |